missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8A Rat anti-Mouse, PerCP-Cy5.5, Clone: 2.43, Abnova™
Rat monoclonal antibody raised against Cd8a.
Brand: Abnova MAB12214.100ug
This item is not returnable.
View return policy
Specifications
| CD8A | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| BB154331/Ly-2/Ly-35/Ly-B/Lyt-2 | |
| Rat | |
| Affinity Chromatography | |
| RUO | |
| 12525 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 2.43 | |
| Rat monoclonal antibody raised against Cd8a. | |
| In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
| Cd8a | |
| Cd8a | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction