missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Rat, V450, Clone: 2.43, enQuireBio™
Rat Monoclonal Antibody
€ 169.05 - € 327.60
Specifications
| Antigen | CD8a |
|---|---|
| Clone | 2.43 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15982288
|
EnquireBio
QAB61-V450-25ug |
25 μg |
€ 169.05
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15972288
|
EnquireBio
QAB61-V450-100ug |
100 μg |
€ 327.60
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD8a | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12525 | |
| IgG2b | |
| Protein A/G Purified |
| 2.43 | |
| Flow Cytometry | |
| Violet Fluor 450 (V450) | |
| RUO | |
| NP_033987 | |
| Cd8a | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title