missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Antibody (479608), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5218B
This item is not returnable.
View return policy
Description
CD9 Monoclonal antibody specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| CD9 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 479608 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| NS0 mouse myeloma cell line transfected with mouse CD9, Pro2-Val226, Accession # P40240 | |
| 0.1 mL | |
| Cancer, Stem Cell Markers | |
| 928 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction