Learn More
CD9 Antibody (479608), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB5218D
Product Code. 18554376
  • € 712.00 / 0.10mL
Description
CD9 Monoclonal antibody specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD9
Monoclonal
DyLight 350
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, Immunohistochemistry, CyTOF
479608
Flow Cytometry, Immunohistochemistry, CyTOF-ready
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
NS0 mouse myeloma cell line transfected with mouse CD9, Pro2-Val226, Accession # P40240
0.1 mL
Cancer, Stem Cell Markers
928
Store at 4°C in the dark.