missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Antibody (MM0180-5H15), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12184AF350
This item is not returnable.
View return policy
Description
CD9 Monoclonal specifically detects CD9 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD9 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD9 | |
| Human recombinant CD9 | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot, Flow Cytometry | |
| MM0180-5H15 | |
| Western Blot, Flow Cytometry | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| Mouse | |
| 0.1 mL | |
| 928 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction