missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Antibody (MM2/57), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28364
This item is not returnable.
View return policy
Description
CD9 Monoclonal specifically detects CD9 in Human, Mouse, Canine, Feline, Hamster, Monkey, Rabbit samples. It is validated for Flow Cytometry.
Specifications
| CD9 | |
| Monoclonal | |
| FITC | |
| P21926 | |
| CD9 | |
| The immunogen is a peptide made to the amino acid region CD9 | |
| 1 mL | |
| Primary | |
| Human, Mouse, Canine, Feline, Hamster, Monkey, Rabbit | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry | |
| MM2/57 | |
| Flow Cytometry 10 μL/10^6 cells | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 928 | |
| Store at 4C. Do not freeze. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction