Learn More
CD9 Mouse anti-Human, FITC, Clone: MM2/57, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-65070-100TESTS
This item is not returnable.
View return policy
Description
The CD9 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD9. This antibody recognizes human antigen. The CD9 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD9 | |
| Monoclonal | |
| 10 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| MM2/57 | |
| FITC | |
| 100 Tests | |
| 928 | |
| IgG2b |