Learn More
CD90.2 Rat, APC, Clone: 30-H12, enQuireBio™
Rat Monoclonal Antibody
€ 131.25 - € 327.60
Specifications
| Antigen | CD90.2 |
|---|---|
| Clone | 30-H12 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15912168
|
EnquireBio
QAB52-APC-25ug |
25 μg |
€ 131.25
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15902168
|
EnquireBio
QAB52-APC-100ug |
100 μg |
€ 327.60
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD90.2 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 21838 | |
| IgG2b | |
| Protein A/G Purified |
| 30-H12 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_033408 | |
| Thy1 | |
| Primary | |
| Store at 2-4°C. |