missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.2 Rat, PerCP-Cy5.5, Clone: 30-H12, enQuireBio™
Rat Monoclonal Antibody
€ 162.00 - € 326.00
Specifications
Antigen | CD90.2 |
---|---|
Clone | 30-H12 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952168
|
EnquireBio
QAB52-PCP55-25UG |
25 μg |
€ 162.00
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942168
|
EnquireBio
QAB52-PCP55-100UG |
100 μg |
€ 326.00
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD90.2 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
21838 | |
IgG2b | |
Protein A/G Purified |
30-H12 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
RUO | |
NP_033408 | |
Thy1 | |
Primary | |
Store at 2-4°C. |