missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
€ 178.00 - € 344.00
Specifications
| Antigen | CD90.2 (Thy-1.2) |
|---|---|
| Clone | 53-2.1 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055567
|
Invitrogen™
M028T03B01-A |
100 μg |
€ 344.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055761
|
Invitrogen™
M028T02B01-A |
25 μg |
€ 178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD90.2 (Thy-1.2) Monoclonal antibody specifically detects CD90.2 (Thy-1.2) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD90.2 (Thy-1.2) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P01831 | |
| 21838 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 53-2.1 | |
| Flow Cytometry | |
| NovaFluor Blue 510 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
| IgG2a κ | |
| Affinity chromatography | |
| Thy1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title