missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD90 Monoclonal Antibody (F15-42-1)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA181930

 View more versions of this product

Product Code. 11560083

  • € 228.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is human brain or thymus. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD90 antibody, clone F15-42-1 recognizes the human CD90 cell surface antigen, a approximately 25 kDa glycoprotein homologous to rat Thy1. Mouse anti Human CD90 antibody, clone F15-42-1 recognizes the human CD90 cell surface antigen, a approximately 25 kDa glycoprotein homologous to rat Thy1.

CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD90
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P04216
Thy1
Purified human brain Thy-1.
20 μg
Primary
Human, Cynomolgus Monkey
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry
F15-42-1
Unconjugated
Thy1
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen
Mouse
Protein A
RUO
102139037, 7070
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.