Learn More
Invitrogen™ CD90 Monoclonal Antibody (F15-42-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516671
Description
The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is human brain or thymus. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD90 antibody, clone F15-42-1 recognizes the human CD90 cell surface antigen, a approximately 25 kDa glycoprotein homologous to rat Thy1. Mouse anti Human CD90 antibody, clone F15-42-1 recognizes the human CD90 cell surface antigen, a approximately 25 kDa glycoprotein homologous to rat Thy1.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
Specifications
| CD90 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P04216 | |
| Thy1 | |
| Purified human brain Thy-1. | |
| 200 μg | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry | |
| F15-42-1 | |
| Unconjugated | |
| Thy1 | |
| CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
| Mouse | |
| Protein A | |
| RUO | |
| 102139037, 7070 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.