missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (1aG4/C5), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-44877V
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse, Human (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CD90/Thy1 | |
| Monoclonal | |
| DyLight 405 | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody 1aG4/C5 recognizes alloantigen Thy-1.2 of mouse Thy-1 (CD90); it does not cross-react with Thy-1.1 alloantigen. Thy-1 is a 18 kDa cell surface GPI linked glycoprotein (Immunoglobulin superfamily); in the mouse it is expressed on all thymocytes and peripheral T lymphocytes. | |
| Store at 4C in the dark. | |
| IgG3 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 1aG4/C5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| THY1 | |
| thymocytes from C3H/Di mice (alelic form: Thy-1.2 positive) | |
| 0.1 mL | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 7070 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction