missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (AF-9), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47755PEATT594
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human, Bovine samples. It is validated for Flow Cytometry.
Specifications
| CD90/Thy1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| THY1 | |
| Human T-lymphoma cells | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 18-35kDa, identified as CD90 (also known as Thy1). CD90 is a member of the immunoglobulin superfamily. It may contribute to inhibition of proliferation/differentiation of hematopoietic stem cells and neuron memory formation in the CNS. It consists of a single Ig domain (112 amino acids; 25-35 kDa) inserted into the cell membrane via a GPI anchor. Expressed by hematopoietic stem cells and neurons in all species studied. Its highly expressed in connective tissue and various fibroblast and stromal cell lines, expressed on all thymocytes and peripheral T cells in mice, but in humans expressed only on small % fetal thymocytes, 10-40% of CD34+ cells in bone marrow, and <1% of CD3+CD4+ lymphocytes in peripheral circulation. It is also expressed by human lymph node HEV endothelium but not other endothelia. Lastly, it is expressed by a limited number of lymphoblastoid and leukemic cell lines. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| AF-9 | |
| Flow Cytometry | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Mouse | |
| Protein A or G purified | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 7070.0 | |
| Human, Bovine | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction