Learn More
CD90/Thy1 Antibody (OX-7), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65543PE
Product Code. 18064585
  • € 618.00 / 0.25mL
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Rat samples. It is validated for Flow Cytometry, Product Image.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD90/Thy1
Monoclonal
PE
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Mouse
Protein A purified
RUO
Primary
CD90
Store at 4C in the dark.
IgG1
Flow Cytometry
OX-7
Flow Cytometry
THY1
Rat Thy 1 antigen
0.25 mL
Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
7070