Learn More
CD93 Mouse anti-Human, BUV563, Clone: R139, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 749194
Product Code. 17210755
  • ONLINE EXCLUSIVE PRICE
    € 540.00
    € 500.00 / 50µg
    Save € 40.00
    7% Off
Description
CD93 Monoclonal antibody specifically detects CD93 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD93
Monoclonal
Brilliant Ultraviolet 563
C1QR1; C1qRP; C1qR(P); C1q/MBL/SPA Receptor; MXRA4; GR11; Dj737e23.1
Clq-CLF-binding proteins
RUO
Human
IgG2b κ
Flow Cytometry
R139
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody.