Learn More
CD93 Mouse anti-Human, BUV563, Clone: R139, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 749194
This item is not returnable.
View return policy
Description
CD93 Monoclonal antibody specifically detects CD93 in Human samples. It is validated for Flow Cytometry
Specifications
| CD93 | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| C1QR1; C1qRP; C1qR(P); C1q/MBL/SPA Receptor; MXRA4; GR11; Dj737e23.1 | |
| Clq-CLF-binding proteins | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| R139 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |