missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Antibody (1032011) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10581JF669
This item is not returnable.
View return policy
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry
Specifications
| CD94 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
| Human embryonic kidney cell HEK293-derived human CD94, Lys32-Ile179, Accession # Q13241.2 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 1032011 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Innate Immunity, Lipid and Metabolism | |
| 3824 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction