missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD94 Antibody (131412), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1058Z
This item is not returnable.
View return policy
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry
Specifications
| CD94 | |
| Monoclonal | |
| DyLight 755 | |
| CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
| BaF3 mouse pro-B cell line transfected with human CD94 and NKG2A | |
| 0.5 mL | |
| Innate Immunity, Lipid and Metabolism | |
| 3824 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 131412 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction