missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD94 Monoclonal Antibody (18d3), PE
Description
CD94 Monoclonal Antibody for Flow
Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | CD94 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 18d3 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with <0.1% sodium azide; pH 7.5 |
| Gene | KLRD1 |
| Gene Accession No. | O54707 |
| Gene Alias | Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?