Learn More
Invitrogen™ CD94 Monoclonal Antibody (1H1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515817
Description
MA5-15817 targets CD94 in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-15817 immunogen is purified recombinant fragment of human CD94 expressed in E. Coli. MA5-15817 detects CD94 which has a predicted molecular weight of approximately 21kDa.
Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| CD94 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q13241 | |
| KLRD1 | |
| Purified recombinant fragment of human CD94 expressed in E. Coli. | |
| RUO | |
| 3824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Western Blot | |
| 1H1 | |
| Unconjugated | |
| KLRD1 | |
| Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.