missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD96 Antibody (051) [FITC], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26893F
This item is not returnable.
View return policy
Description
CD96 Monoclonal antibody specifically detects CD96 in Mouse samples. It is validated for ELISA
Specifications
| CD96 | |
| Monoclonal | |
| FITC | |
| CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD96 (Uniprot#: Q3U0X8; Met1-Met536). | |
| 0.1 mL | |
| Immunology | |
| 10225 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 051 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction