missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD96 Monoclonal Antibody (630612)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524281
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.
Specifications
| CD96 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q3U0X8 | |
| Cd96 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD96 Val22-Met536. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 630612 | |
| Unconjugated | |
| Cd96 | |
| 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile | |
| Rat | |
| Protein A/G | |
| RUO | |
| 84544 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction