missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD96 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547823
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 3% cross-reactivity with recombinant mouse CD96 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.
Specifications
| CD96 | |
| Polyclonal | |
| Unconjugated | |
| Cd96 | |
| 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 10225 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P40200 | |
| Cd96 | |
| Mouse myeloma cell line NS0-derived recombinant human CD96 Lys25-Asp501. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction