missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Antibody (3A7), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | CD97 |
|---|---|
| Clone | 3A7 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200 |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043295
|
Novus Biologicals
NBP3-26673-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043868
|
Novus Biologicals
NBP3-26673-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD97 Monoclonal antibody specifically detects CD97 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| CD97 | |
| ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
| Synthesized peptide [UniProt P48960] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3A7 | |
| ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| GPCR, Immunology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 976 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title