missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99507B
This item is not returnable.
View return policy
Description
CD97 Polyclonal antibody specifically detects CD97 in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CD97 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Biotin | |
| CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CD97 | |
| 0.1 mL | |
| GPCR, Immunology | |
| 976 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction