missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Isoform 1 Antibody (380903), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2529PEATT594
This item is not returnable.
View return policy
Description
CD97 Isoform 1 Monoclonal antibody specifically detects CD97 Isoform 1 in Human samples. It is validated for Flow Cytometry
Specifications
| CD97 Isoform 1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 976 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 380903 | |
| Flow Cytometry | |
| CD97, TM7LN1 | |
| Mouse myeloma cell line NS0-derived recombinant human CD97, Gln21-Gln398, Accession # NP_001775.2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction