missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Isoform 1 Antibody (587702) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3734JF585
This item is not returnable.
View return policy
Description
CD97 Isoform 1 Monoclonal antibody specifically detects CD97 Isoform 1 in Mouse samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,CyTOF
Specifications
| CD97 Isoform 1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD97, TM7LN1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD97v2, Gln24-His384, Accession # AAH06676 | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 587702 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 976 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction