missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Polyclonal Antibody

CD97 Polyclonal Antibody
Brand: Invitrogen PA5103014
This item is not returnable.
View return policy
Description
CD97 Polyclonal Antibody for Western Blot
Specifications
| CD97 | |
| Polyclonal | |
| Unconjugated | |
| ADGRE5 | |
| AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 976 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48960 | |
| ADGRE5, CD97 | |
| A synthesized peptide derived from human CD97(Accession P48960), corresponding to amino acid residues S451-D501. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction