missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD98 Antibody (590559) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5920JF585
This item is not returnable.
View return policy
Description
CD98 Monoclonal antibody specifically detects CD98 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| CD98 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD98, Asp105-Gln529, Accession # P08195 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 590559 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| 6520 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction