missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD98 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596401

Product Code. 16539241

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RAPRCRELPA QKWWHTGALY RIGDLQAFQG HGAGNLAGLK GRLDYLSSLK VKGLVLGPIH KNQKDDVAQT DLLQIDPNFG SKEDFDSLLQ SAKKKSIRVI LDLTPNYRGE NSWFSTQVDT VATKVKDALE FWLQAGVDGF QVRDIENLKD ASSFLAEWQN ITKGFSEDRL LIAGTNSSDL QQILSLLESN KDLLLTSSYL SDSGSTGEHT KSLVTQYLNA TGNRWCSWSL SQARLLTSFL PAQLLRLYQL MLFTLPGTPV FSYGDEIGLD AAALPGQPME APVMLWDESS FPDIPGAVSA NMTVKGQSED PGSLLSLFRR LSDQRSKERS LLHGDFHAFS AGPGLFSYIR HWDQNERFLV VLNFGDVGLS AGLQASDLPA SASLPAKADL LLSTQPGREE GSPLELERLK LEPHEGLLLR FPYAA Positive Samples: 293T, MCF7, SW480, SKOV3, Mouse kidney, Mouse lung, Mouse testis, Mouse ovary; Cellular Location: Apical cell membrane, Melanosome, Single-pass type II membrane protein.

CD98 (4F2) is a type II transmembrane glycoprotein which serves as the heavy chain of the heterodimeric amino acid transporters (HATs). CD98, linked to various light chains by disulfide bond, is responsible for cell surface expression and basolateral localization of this transporter complex in polarized epithelial cells and also interacts with beta1 integrins and increases their affinity for ligand. Besides its roles in amino acid transport, CD98 is thus involved in cell fusion and activation. It is implicated in regulation of cellular differentiation, growth and apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD98
Polyclonal
Unconjugated
SLC3A2
4F2; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2hc; 4T2HC; AI314110; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; Cd98; CD98 antigen; CD98 heavy chain; CD98HC; Ly10; Ly-10; Ly-m10; Lymphocyte activation antigen 4F2 large subunit; MDU1; Mgp-2hc; monoclonal antibody 44D7; NACAE; RGD:3073}; SLC3A2; slc3a2 {ECO:0000312; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; solute carrier family 3 member 2; solute carrier family 3, member 2; type II transmembrane protein
Rabbit
Affinity Chromatography
RUO
17254, 50567, 6520
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.35 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
P08195, P10852, Q794F9
SLC3A2
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.