Learn More
CD99 Antibody (CL12929) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18550MFV450
Product Code. 30293709
  • € 791.00 / 0.10mL
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD99
Monoclonal
mFluor Violet 450 SE
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
This antibody was developed using a synthetic peptide derived from P14209, with the exact immunogen sequence remaining proprietary.
0.1 mL
Immunology
4267
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12929
50mM Sodium Borate
Mouse
Protein A purified