Learn More
CD99 Antibody (MIC2/8119R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21121AF405
Code produit. 18520826
  • € 726.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CD99
Monoclonal
Alexa Fluor 405
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
Recombinant fragment (around aa1-185) of human CD99 protein (exact sequence is proprietary)
0.1 mL
Immunology
4267
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
MIC2/8119R
50 mM Sodium Borate