missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (MIC2/8119R), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21121JF549
This item is not returnable.
View return policy
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD99 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Recombinant fragment (around aa1-185) of human CD99 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology | |
| 4267 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MIC2/8119R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction