missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (rMIC2/8497), Novus Biologicals™
Mouse Monoclonal Antibody
€ 249.00 - € 494.00
Specifications
| Antigen | CD99 |
|---|---|
| Clone | rMIC2/8497 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618014
|
Novus Biologicals
NBP3-20463-100ug |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620855
|
Novus Biologicals
NBP3-20463-20ug |
20 μg |
€ 249.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD99 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| 10mM PBS with 0.05% BSA | |
| 4267 | |
| IgM | |
| Protein A purified |
| rMIC2/8497 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Recombinant fragment (around aa1-185) of human CD99 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title