Learn More
CD99-L2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98287JF646
This item is not returnable.
View return policy
Description
CD99-L2 Polyclonal antibody specifically detects CD99-L2 in Mouse samples. It is validated for ELISA
Specifications
| CD99-L2 | |
| Produced in rabbits immunized with purified, recombinant Mouse CD99-L2 (Accession#: NP_612182.1; Met 1-Ala 164) | |
| Mouse | |
| Purified |
| Polyclonal | |
| RUO | |
| Store at 4C in the dark. | |
| IgG |