missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Monoclonal Antibody (OTI2H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800821S
This item is not returnable.
View return policy
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Western BlotSpecifications
| CD99 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14209 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4267 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2H6 | |
| Unconjugated | |
| CD99 | |
| 1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2 | |
| Full length human recombinant protein of human CD99 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction