missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Monoclonal Antibody (OTI5C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800904
This item is not returnable.
View return policy
Description
CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.Specifications
| CD99 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14209 | |
| CD99 | |
| Full length human recombit protein of human CD99 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C8 | |
| Unconjugated | |
| CD99 | |
| HBA71, MIC2, MIC2X, MIC2Y, MSK5X | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4267 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction