missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD99L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598691
This item is not returnable.
View return policy
Description
CD99L2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
CD99L2 may function as a homophilic adhesion molecule. CD99L2 functions in leukocyte-endothelial cell interactions during leukocyte extravasation, and in particular, at the diapedesis step. CD99L2 does not seem to be involved in docking of leukocytes to the vessel wall or in lymphocyte diapedesis.
Specifications
| CD99L2 | |
| Polyclonal | |
| Unconjugated | |
| Cd99l2 | |
| AW548191; CD99; Cd99 antigen-like 2; CD99 antigen-like protein 2; CD99 molecule like 2; CD99 molecule-like 2; CD99B; CD99L2; MIC2 (monoclonal Imperial Cancer Research Fund 2)-like 1; MIC2 like 1; MIC2 like-1; MIC2L1; MIC2-like protein 1; protein rhombex-40; Rhombencephalic expression protein 40 kDa; Rhombex40; Rhombex-40; single-pass transmembrane protein; UNQ1964/PRO4486; vms-tm2; Xap89 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 83692 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TCZ2 | |
| Cd99l2 | |
| Recombinant Human CD99 antigen-like protein 2 protein (26-145AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction