missing translation for 'onlineSavingsMsg'
Learn More

CDA Monoclonal Antibody (921CT14.2.1)
GREENER_CHOICE

Product Code. 17197208 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197208 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17197208 Supplier Thermo Scientific Supplier No. MA537625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CDA Monoclonal Antibody (921CT14.2.1)

CDA Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDA
Applications Western Blot
Classification Monoclonal
Clone 921CT14.2.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CDA
Gene Accession No. P32320
Gene Alias 2210401N16Rik; CDA; CDD; cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; EC 3.5.4.5; small cytidine deaminase
Gene Symbols CDA
Host Species Mouse
Immunogen purified His-tagged CDA protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.