missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589551
This item is not returnable.
View return policy
Description
Immunogen sequence: MAQKRPACTL KPECVQQLLV CSQEAKKSAY CPYSHFPVGA ALLTQEGRIF KGCNIENACY PLGICAERTA IQKAVSEGYK DFRAIAIASD MQDDFISPCG ACRQVMREFG TNWPVYMTKP DGTYIVMTVQ ELLPSSFGPE DLQKTQ; Positive Samples: HeLa, HepG2, 293T, Jurkat, SW620, Mouse kidney.
This gene encodes an enzyme involved in pyrimidine salvaging. The encoded protein forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. It is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in this gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.
Specifications
| CDA | |
| Polyclonal | |
| Unconjugated | |
| CDA | |
| 2210401N16Rik; CDA; CDD; cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; EC 3.5.4.5; small cytidine deaminase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 72269, 978 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32320, P56389 | |
| CDA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-146 of human CDA (NP_0017761). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction