missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDADC1 Antibody (2E8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00081602-M02
This item is not returnable.
View return policy
Description
CDADC1 Monoclonal antibody specifically detects CDADC1 in Human, Rat samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| CDADC1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| bA103J18.1, cytidine and dCMP deaminase domain containing 1, cytidine and dCMP deaminase domain-containing protein 1, MGC150615, MGC41774, MGC57136, NYD-SP15, protein kinase NYD-SP15, Testis development protein NYD-SP15 | |
| CDADC1 (NP_112173, 423 a.a. ∽ 514 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KCPCDECVPLIKGAGIKQIYAGDVDVGKKKADISYMRFGELEGVSKFTWQLNPSGAYGLEQNEPERRENGVLRPVPQKEEQHQDKKLRLGIH | |
| 0.1 mg | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 2E8 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_112173 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 81602 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction