missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDAN1 Antibody (Cod177) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 386.00
Specifications
| Antigen | CDAN1 |
|---|---|
| Clone | Cod177 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649046
|
Novus Biologicals
NBP2-50176-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657917
|
Novus Biologicals
NBP2-50176 |
0.1 mL |
€ 386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDAN1 Monoclonal antibody specifically detects CDAN1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CDAN1 | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology | |
| PBS | |
| 146059 | |
| IgG2b | |
| Protein A purified |
| Cod177 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CDA1, CDAI, CDA-I, codanin, codanin-1, congenital dyserythropoietic anemia, type I, discs lost homolog, DLT, PRO1295 | |
| aa 294-607 of cloned human protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title