missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC123 Antibody (OTI1F8), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71391DL594
This item is not returnable.
View return policy
Description
CDC123 Monoclonal antibody specifically detects CDC123 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| CDC123 | |
| Monoclonal | |
| DyLight 594 | |
| cell division cycle 123 homolog (S. cerevisiae), cell division cycle protein 123 homolog, chromosome 10 open reading frame 7, D123C10orf7, FLJ13863, HT-1080, Protein D123, PZ32 | |
| Full length recombinant protein of human CDC123 (NP_006014) produced in HEK293T cells. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8872 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction