missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC123 Monoclonal Antibody (OTI1E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505651S
This item is not returnable.
View return policy
Description
CDC123 Monoclonal antibody specifically detects CDC123 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CDC123 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75794 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8872 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E12 | |
| Unconjugated | |
| CDC123 | |
| AA959893; C10orf7; Cdc123; cell division cycle 123; cell division cycle 123 homolog; cell division cycle protein 123 homolog; Cell division cycle protein 123-like protein; D123; D123 gene product; G431001I09Rik; HT-1080; hypothetical protein; Protein D123; PZ32; QccE-12339 | |
| Full length human recombinant protein of human CDC123 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction