Learn More
Cdc13 Antibody (6F11/2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NB200-576
Product Code. 18755053
  • € 372.00 / 0.20mg
Description
Cdc13 Monoclonal specifically detects Cdc13 in S. pombe, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
0.2 mg
IgG2a
Full length cdc13