missing translation for 'onlineSavingsMsg'
Learn More

CDC16 Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30511772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30511772 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511772 Supplier Novus Biologicals Supplier No. NBP338396PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC16 Polyclonal antibody specifically detects CDC16 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC16
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias ANAPC6subunit 6, CDC16 (cell division cycle 16, S. cerevisiae, homolog), CDC16 cell division cycle 16 homolog (S. cerevisiae), CDC16 homolog, CDC16Hs, cell division cycle 16 homolog (S. cerevisiae), cell division cycle protein 16 homolog, Cyclosome subunit 6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 371-620 of human CDC16 (NP_003894.3).,, Sequence:, MLYIGLEYGLTNNSKLAERFFSQALSIAPEDPFVMHEVGVVAFQNGEWKTAEKWFLDALEKIKAIGNEVTVDKWEPLLNNLGHVCRKLKKYAEALDYHRQALVLIPQNASTYSAIGYIHSLMGNFENAVDYFHTALGLRRDDTFSVTMLGHCIEMYIGDSEAYIGADIKDKLKCYDFDVHTMKTLKNIISPPWDFREFEVEKQTAEETGLTPLETSRKTPDSRPSLEETFEIEMNESDMMLETSMSDHST
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8881
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.