missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDC16 Antibody (SR3221), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 316.00 - € 382.00
Specifications
| Antigen | CDC16 |
|---|---|
| Clone | SR3221 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041583
|
Novus Biologicals™
NBP3-27756-50ul |
50 μL |
€ 316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041754
|
Novus Biologicals™
NBP3-27756-100ul |
100 μL |
€ 382.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC16 Monoclonal antibody specifically detects CDC16 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| CDC16 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ANAPC6subunit 6, CDC16 (cell division cycle 16, S. cerevisiae, homolog), CDC16 cell division cycle 16 homolog (S. cerevisiae), CDC16 homolog, CDC16Hs, cell division cycle 16 homolog (S. cerevisiae), cell division cycle protein 16 homolog, Cyclosome subunit 6 | |
| A synthesized peptide derived from human CDC16 (Uniprot #: Q13042) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3221 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 8881 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title