missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC2/CDK1 Antibody (Y100.4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2716G
This item is not returnable.
View return policy
Description
CDC2/CDK1 Monoclonal specifically detects CDC2/CDK1 in S. pombe samples. It is validated for Western Blot.
Specifications
| CDC2/CDK1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDK1 | |
| Peptide spanning the C-terminal 14 residues of S. pombe CDC2/CDK1 | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| 983 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot | |
| Y100.4 | |
| Western Blot | |
| CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| S. pombe | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction