missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc20, p Thr79 Antibody (BT2.1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50246AF488
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cdc20 Monoclonal specifically detects Cdc20 in Xenopus samples. It is validated for Western Blot, ELISA.
Specifications
| Cdc20 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDC20 | |
| GPKMQGpTPSRAG. pT represents phosphorylated Thr79. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 991 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| BT2.1 | |
| Western Blot, ELISA | |
| CDC20 cell division cycle 20 homolog, CDC20 cell division cycle 20 homolog (S. cerevisiae), cell division cycle 20 homolog (S. cerevisiae), cell division cycle protein 20 homolog, MGC102824, p55CDCS. cerevisiae, homolog) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction