Learn More
CDC25A Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-213IR
This item is not returnable.
View return policy
Description
CDC25A Polyclonal antibody specifically detects CDC25A in Human samples. It is validated for Western Blot
Specifications
| Polyclonal | |
| 0.1 mL | |
| Purified |
| DyLight 755 | |
| Store at 4C in the dark. |